Potassium bromide, crystal rectangle for spectrophotometry, polished

Supplier: INTERNATIONAL CRYSTAL LABS MS
Warning

0002C-86R 0002C-99 0002C-75 0002C-168 0002C-69
100211-842EA 176.85 USD
100211-842 100214-558 100214-550 100213-328 100214-022
Potassium bromide, crystal rectangle for spectrophotometry, polished
Potassium bromide

Potassium Bromide is commonly used for infrared transmission windows in gas and liquid sample cells used with infrared and FTIR spectrophotometers and for beam splitters for spectrophotometers. KBr windows are soft and hygroscopic. ICL is the only company

Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR