Sodium chloride, crystal disk for spectroscopy, for CO₂ lasers, polished, drilled

Supplier: INTERNATIONAL CRYSTAL LABS MS

Synonyms: Saline, NaCl, Sea salt, Halite

0002A-84
100212-706EA 88.57 USD
100212-706
Sodium chloride, crystal disk for spectroscopy, for CO₂ lasers, polished, drilled
Sodium chloride

Quality crystal optics for spectroscopy and CO2 lasers.

Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR