Calcium fluoride, crystal random cuttings for spectrophotometry

Supplier: INTERNATIONAL CRYSTAL LABS MS
Warning

0002-4503
100212-690EA 1191.15 USD
100212-690
Calcium fluoride, crystal random cuttings for spectrophotometry
Calcium fluoride

International Crystal Labs celebrated its 40th anniversary in 2002. They have produced quality crystal optics for spectroscopy and CO2 lasers since 1962. ICL operates a vertically integrated crystal optics production plant and grow their own crystals.

Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: 2500 °C (1013 hPa)
Melting Pt: 1418 °C
Density: 3.18 g/cm³ (20 °C)
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR