Fluorometry Disposable Cuvettes, Polymethyl Metacrylate (PMMA), International Crystal Laboratories
Supplier: International Crystal Labs
0023-7477
100212-658PK 191.46 USD
100212-658
Fluorometry Disposable Cuvettes, Polymethyl Metacrylate (PMMA), International Crystal Laboratories
Cuvettes Spectrophotometer Cuvettes
These polymethyl methacrylate cuvettes offer superior optical performance coupled with the convenience of being disposable
  • Superior optical performance
  • Disposable
Using disposable cuvettes eliminates cleaning and cross contamination.