Potassium bromide, crystal disk for spectrophotometry, polished, drilled

Supplier: INTERNATIONAL CRYSTAL LABS MS
Warning

0002C-118
100212-568EA 96.62 USD
100212-568
Potassium bromide, crystal disk for spectrophotometry, polished, drilled
Potassium bromide

Potassium Bromide is commonly used for infrared transmission windows in gas and liquid sample cells used with infrared and FTIR spectrophotometers and for beam splitters for spectrophotometers. KBr windows are soft and hygroscopic. ICL is the only company

Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435 °C (1013 hPa)
Melting Pt: 730 °C
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR