Potassium chloride, crystal disk for spectrophotometry, polished

Supplier: INTERNATIONAL CRYSTAL LABS MS

Synonyms: Potassiumchloride

0002B-125 0002B-1424 0002B-306 0002B-1004 0002B-136 0002B-164 0002B-4548 0002B-115
100215-410EA 69.28 USD
100215-410 100211-348 100215-840 100215-868 100213-320 100211-846 100212-536 100213-310
Potassium chloride, crystal disk for spectrophotometry, polished
Potassium chloride

Potassium Chloride (KCl) is commonly used for infrared transmission crystal windows in gas and liquid sample cells used with infrared and FTIR spectrophotometers. KCl crystal windows are particularly useful for spatter barrier windows in CO2 lasers becaus

Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR