Zinc selenide, crystal disk for spectrophotometry, polished

Supplier: INTERNATIONAL CRYSTAL LABS MS
Danger

Synonyms: Zinc (II) selenide

0002J-1823 0002J-1831 0002J-1829 0002J-1822 0002J-1825 0002J-1826 0002J-1828 0002J-1821 0002J-4464
100211-392EA 380 USD
100211-392 100214-650 100215-494 100211-630 100216-048 100216-094 100215-492 100211-718 100211-642
Zinc selenide, crystal disk for spectrophotometry, polished
Zinc selenide

Zinc Sulfide (ZnS) is used as a transmission window for IR spectroscopy. Zinc Sulfide (ZnS) is or has been produced under the trademarks Cleartran® and Irtran® .

Formula: SeZn
MW: 144.35 g/mol
Melting Pt: 1793
Density: 5.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1 III
Merck Index: 12,10290

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR