Sodium chloride, crystal disk for spectroscopy, for CO₂ lasers, polished

Supplier: INTERNATIONAL CRYSTAL LABS MS

Synonyms: Saline, NaCl, Sea salt, Halite

0002A-82 0002A-83 0002A-1003 0002A-68 0002A-110 0002A-330 0002A-67 0002A-623
100211-330EA 58.75 USD
100211-330 100215-828 100211-514 100213-306 100216-108 100216-118 100211-328 100211-868
Sodium chloride, crystal disk for spectroscopy, for CO₂ lasers, polished
Sodium chloride

Quality crystal optics for spectroscopy and CO2 lasers.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR