Potassium bromide, crystal disk for spectrophotometry, polished

Supplier: INTERNATIONAL CRYSTAL LABS MS
Warning

0002C-1854 0002C-175 0002C-248 0002C-244 0002C-1005 0002C-4085 0002C-74 0002C-144 0002C-81
100211-534EA 72.49 USD
100211-534 100215-604 100215-836 100213-338 100213-474 100211-548 100214-548 100211-494 100214-552
Potassium bromide, crystal disk for spectrophotometry, polished
Potassium bromide

Potassium Bromide is commonly used for infrared transmission windows in gas and liquid sample cells used with infrared and FTIR spectrophotometers and for beam splitters for spectrophotometers. KBr windows are soft and hygroscopic. ICL is the only company

Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR