Ytterbium ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS
Warning

93-7035-50X50MM 93-7035-25X25MM
100208-592EA 1220.06 USD
100208-592 100211-290
Ytterbium ≥99.9%, foil
Ytterbium
Formula: Yb
Boiling Pt: 1427 °C
Melting Pt: 824 °C
Density: 6.97
MDL Number: MFCD00011286
CAS Number: 7440-64-4
EINECS: 231-173-2
Merck Index: 12,10239

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR