Neodymium(III) oxide ≥99.9% (Nd basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide
93-6031-250G
93-6031-50G
100208-730EA
469.91
USD
100208-730
100211-282
Neodymium(III) oxide ≥99.9% (Nd basis)
Neodymium (III) oxide
Formula:
Nd₂O₃ Boiling Pt: 3760 °C (1013 hPa) Melting Pt: 2272 °C Density: 7.24 |
MDL Number:
MFCD00011134 CAS Number: 1313-97-9 EINECS: 215-214-1 Merck Index: 14,06450 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...