Sulfur ≥99.999%

Supplier: STREM CHEMICALS INC MS
Warning

93-1616-500G 93-1616-25G 93-1616-100G
100208-126EA 1293.66 USD
100208-126 100197-720 100211-224
Sulfur ≥99.999%
Sulphur
Formula: S
Boiling Pt: 444.67 °C
Melting Pt: 112.8 °C
Density: 2.07
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1,III
Merck Index: 13,09059

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR