Thallium(I) trifluoromethanesulfonate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: (Thallium triflate)

81-8500-1G 81-8500-25G 81-8500-5G
100211-180EA 60.86 USD
100211-180 100211-182 100211-184
Thallium(I) trifluoromethanesulfonate ≥99%
Thallium(I) trifluoromethanesulfonate

Metal Triflate

MDL Number: MFCD00061608
CAS Number: 73491-36-8
UN: 1707
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR