Vinyltriethylsilane(hexafluoroacetylacetonato)silver(I) ≥99.9% (Ag basis)

Supplier: STREM CHEMICALS INC MS
Warning

47-8000-5G 47-8000-1G
100211-128EA 314.22 USD
100211-128 100211-126
Vinyltriethylsilane(hexafluoroacetylacetonato)silver(I) ≥99.9% (Ag basis)
Vinyltriethylsilane(hexafluoroacetylacetonato)silver(I)

Metal Beta-diketonates, Volatile Precursors for CVD

MDL Number: MFCD03094581
CAS Number: 177279-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR