Bis(cyclopentadienyl)ruthenium ≥99% (99.9%-Ru)

Supplier: STREM CHEMICALS INC MS
Warning

44-6200-1G 44-6200-5G
100211-100EA 176.92 USD
100211-100 100211-102
Bis(cyclopentadienyl)ruthenium ≥99% (99.9%-Ru)
Bis(cyclopentadienyl)ruthenium

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₀Ru
Melting Pt: 194…198 °C
MDL Number: MFCD00001442
CAS Number: 1287-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR