Strontium carbonate ≥99.995%, PURATREM
Supplier: STREM CHEMICALS INC MS
38-1100-50G
38-1100-10G
100211-076EA
401.97
USD
100211-076
100211-074
Strontium carbonate ≥99.995%, PURATREM
Strontium carbonate
Formula:
CO₃Sr Boiling Pt: 2647 °C (1 mmHg) Melting Pt: 1497 °C Density: 3.70 Storage Temperature: Ambient |
MDL Number:
MFCD00011250 CAS Number: 1633-05-2 EINECS: 216-643-7 Merck Index: 14,08838 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...