Aluminium oxide, γ-phase ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

13-2525-2000G 13-2525-500G
100210-984EA 186.83 USD
100210-984 100197-368
Aluminium oxide, γ-phase ≥97%
Aluminium oxide, γ-phase
Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR