4-Carboxyphenylboronic acid pinacol ester ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

05-1037-5G 05-1037-1G
100210-938EA 256.18 USD
100210-938 100210-936
4-Carboxyphenylboronic acid pinacol ester ≥97%
4-Carboxyphenylboronic acid pinacol ester

Boronic Acid and Ester

Formula: C₁₃H₁₇BO₄
Melting Pt: 229…231 °C
MDL Number: MFCD01863710
CAS Number: 180516-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR