Copper(II) chloride dihydrate ≥99% ACS
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Copper dichloride dihydrate, Cupric chloride dihydrate
93-2913-1000G
93-2913-250G
100210-866EA
468.49
USD
100210-866
100209-430
Copper(II) chloride dihydrate ≥99% ACS
Copper (II) chloride dihydrate
Formula:
CuCl₂·2H₂O Melting Pt: 100 °C Density: 2.54 Storage Temperature: Ambient |
MDL Number:
MFCD00149674 CAS Number: 10125-13-0 EINECS: 231-210-2 UN: 2802 ADR: 8,III Merck Index: 13,02660 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...