Diammonium tetrachloropalladate ≥99.998% (Pd basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Ammonium tetrachloropalladate(II, Ammonium tetrachloropalladate(II), Ammonium palladium (II) chloride, Ammonium tetrachloropalladate (II)
02-1275-1G
02-1275-5G
100197-278EA
169.85
USD
100197-278
100210-770
Diammonium tetrachloropalladate ≥99.998% (Pd basis), PURATREM
Diammonium tetrachloropalladate
Formula:
H₈Cl₄N₂Pd Melting Pt: Dec. Density: 2.17 |
MDL Number:
MFCD00010884 CAS Number: 13820-40-1 EINECS: 237-498-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...