Platinum ≥99.999%, powder

Supplier: STREM CHEMICALS INC MS
Warning

78-1800-2G 78-1800-500MG
100210-750EA 778.46 USD
100210-750 100206-836
Platinum ≥99.999%, powder
Platinum
Formula: Pt
Boiling Pt: 3827 °C
Melting Pt: 1769 °C
Density: 21.45
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR