2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-0433-5G 15-0433-1G 15-0433-25G
100210-724EA 169.85 USD
100210-724 100198-616 100202-956
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

BINAP, Phosphine

Formula: C₄₄H₃₂P₂
MDL Number: MFCD00010805
CAS Number: 98327-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR