Europium(III) carbonate hydrate ≥99.9% (Eu basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6304-5G 93-6304-1G
100210-704EA 274.58 USD
100210-704 100205-714
Europium(III) carbonate hydrate ≥99.9% (Eu basis)
Europium(III) carbonate hydrate
Formula: Eu₂(CO₃)₃·xH₂O
MDL Number: MFCD00149702
CAS Number: 86546-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR