2,6-Diisopropylphenylimidoneophylidene molybdenum(VI) bis(t-butoxide)

Supplier: STREM CHEMICALS INC MS
Warning

42-1200-500MG 42-1200-100MG
100210-646EA 839.32 USD
100210-646 100209-930
2,6-Diisopropylphenylimidoneophylidene molybdenum(VI) bis(t-butoxide)
2,6-Diisopropylphenylimidoneophylidene molybdenum(VI) bis(t-butoxide)

Alkoxide, Schrock's Catalyst

MDL Number: MFCD00142847
CAS Number: 126949-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR