Tungstic(VI) acid ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Dihydrogen wolframate, Tungstic acid

93-7410-250G 93-7410-50G
100210-554EA 243.45 USD
100210-554 100196-824
Tungstic(VI) acid ≥99%
Tungstic (VI) acid
Formula: H₂O₄W
Melting Pt: 100 °C (dec.)
Density: 5,5
MDL Number: MFCD00011336
CAS Number: 7783-03-1
EINECS: 231-975-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR