Holmium(III) oxide ≥99.9% (Ho basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Diholmium trioxide, Holmium oxide

93-6713-5G 93-6713-25G
100210-532EA 67.94 USD
100210-532 100207-890
Holmium(III) oxide ≥99.9% (Ho basis)
Holmium (III) oxide
Formula: Ho₂O₃
Boiling Pt: 2600 °C (1013 hPa)
Melting Pt: 2360 °C
Density: 8.36
MDL Number: MFCD00011053
CAS Number: 12055-62-8
EINECS: 235-015-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR