Molybdenum diselenide ≥99.9% (Mo basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-4211-25G 93-4211-5G
100210-406EA 475.57 USD
100210-406 100210-408
Molybdenum diselenide ≥99.9% (Mo basis)
Molybdenum diselenide
Formula: MoSe₂
Melting Pt: > 1200 °C
Density: 6.0
MDL Number: MFCD00014216
CAS Number: 12058-18-3
EINECS: 235-027-9
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR