Germanium ≥99.999%, pieces
Supplier: STREM CHEMICALS INC MS
93-3232-50G
93-3232-10G
100210-368EA
1078.52
USD
100210-368
100210-366
Germanium ≥99.999%, pieces
Germanium
Formula:
Ge Boiling Pt: 2830 °C Melting Pt: 937.4 °C Density: 5.323 Storage Temperature: Ambient |
MDL Number:
MFCD00085310 CAS Number: 7440-56-4 EINECS: 231-164-3 Merck Index: 12,04415 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...