Cobalt(II) fluoride, anhydrous ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cobalt difluoride, Cobaltous fluoride

93-2738-50G 93-2738-10G
100210-318EA 237.78 USD
100210-318 100200-098
Cobalt(II) fluoride, anhydrous ≥99%
Cobalt (II) fluoride
Formula: CoF₂
Boiling Pt: 1400 °C
Melting Pt: ∼1200 °C
Density: 4.46
MDL Number: MFCD00010941
CAS Number: 10026-17-2
EINECS: 233-061-9
UN: 2923
ADR: 8,II
Merck Index: 13,02465

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR