Cobalt(II) phosphate hydrate

Supplier: STREM CHEMICALS INC MS

93-2713-250G 93-2713-50G
100210-310EA 503.88 USD
100210-310 100210-312
Cobalt(II) phosphate hydrate
Cobalt(II) phosphate hydrate
Formula: Co₃(PO₄)₂·xH₂O
Density: 2.769
MDL Number: MFCD00150916
CAS Number: 10294-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR