Calcium ≥99.5%, granules

Supplier: STREM CHEMICALS INC MS
Danger

93-2075-100G 93-2075-500G
100210-248EA 107.57 USD
100210-248 100203-844
Calcium ≥99.5%, granules
Calcium
Formula: Ca
Boiling Pt: 1484 °C
Melting Pt: 842 °C
Density: 1.54
Storage Temperature: Ambient
MDL Number: MFCD00085314
CAS Number: 7440-70-2
EINECS: 231-179-5
UN: 1401
ADR: 4.3,II
Merck Index: 13,01644

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR