Silicon nitride ≥99% (Si basis) < 5 µm

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Silicon(IV) nitride

93-1442-50G 93-1442-250G
100196-508EA 67.94 USD
100196-508 100210-196
Silicon nitride ≥99% (Si basis) < 5 µm
Silicon nitride
Formula: N₄Si₃
Melting Pt: 1900 °C
Density: 3,44
MDL Number: MFCD00011230
CAS Number: 12033-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR