Trichloro(phenyl)silane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Phenyltrichlorosilane

93-1419-100G 93-1419-500G
100198-986EA 48.12 USD
100198-986 100210-190
Trichloro(phenyl)silane ≥97%
Trichloro(phenyl)silane
Formula: C₆H₅Cl₃Si
Boiling Pt: 201 °C
Melting Pt: –40 °C
Density: 1.321
Flash Pt: 196 °F
MDL Number: MFCD00000479
CAS Number: 98-13-5
EINECS: 202-640-8
UN: 1804
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR