Lithium acetate dihydrate ≥98% (99.9%-Li)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Acetic acid lithium salt

93-0362-250G 93-0362-1000G
100210-122EA 117.48 USD
100210-122 100202-518
Lithium acetate dihydrate ≥98% (99.9%-Li)
Lithium acetate dihydrate
Formula: H₃CCOOLi·2H₂O
Melting Pt: Dec.
Density: 1.3
Storage Temperature: Ambient
MDL Number: MFCD00066949
CAS Number: 6108-17-4
EINECS: 208-914-3
Merck Index: 13,05543

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR