Lead(II) citrate trihydrate ≥97%

Supplier: STREM CHEMICALS INC MS

82-0600-100G 82-0600-25G
100198-918EA 142.95 USD
100198-918 100210-102
Lead(II) citrate trihydrate ≥97%
Lead(II) citrate trihydrate
Formula: C₁₂H₁₆O₁₇Pb₃
MDL Number: MFCD00150456
CAS Number: 6107-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR