Lanthanum(III) fluoride, anhydrous ≥99.99% (La basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Lanthanum trifluoride

57-2000-100G 57-2000-25G
100210-022EA 481.23 USD
100210-022 100210-024
Lanthanum(III) fluoride, anhydrous ≥99.99% (La basis), PURATREM
Lanthanum (III) fluoride
Formula: F₃La
Boiling Pt: ∼2327 °C (1013 hPa)
Melting Pt: 1493 °C
Density: 5.936
Storage Temperature: Ambient
MDL Number: MFCD00011073
CAS Number: 13709-38-1
EINECS: 237-252-8
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR