CANNON® CL Low Temperature Viscosity Standards, Cannon Instruments
Supplier: CANNON INSTRUMENT CO
9727-N02.016 9727-N02.032 9727-N02.128 9727-N02.5 9727-N04.016 9727-N04.032 9727-N04.128 9727-N04.5 9727-N06.016 9727-N06.032 9727-N06.128 9727-N06.5 9727-N07.016 9727-N07.032 9727-N07.128 9727-N07.5 9727-N08.016 9727-N08.032 9727-N08.128 9727-N08.5 9727-N09.128 9727-N09.5 9727-N10.016 9727-N10.032 9727-N10.128 9727-N10.5 9727-N11.016 9727-N11.032 9727-N11.128 9727-N11.5 9727-N12.016 9727-N12.032 9727-N12.128 9727-N12.5 9727-N13.016 9727-N13.032 9727-N13.128 9727-N13.5 9727-N16.032 9727-N16.128 9727-N16.5 9727-N18.016 9727-N18.032 9727-N18.128 9727-N18.5 9727-N20.016 9727-N20.032 9727-N20.128 9727-N20.5 9727-N22.016 9727-N22.032 9727-N22.128 9727-N22.5 9727-N24.016 9727-N24.032 9727-N24.128 9727-N24.5 9727-N26.016 9727-N26.032 9727-N26.128 9727-N26.5 9727-N28.016 9727-N28.032 9727-N28.128 9727-N28.5 9727-N30.016 9727-N31.016 9727-N31.032 9727-N31.128 9727-N31.5 9727-N32.016 9727-N32.032 9727-N32.128 9727-N32.5 9727-N33.016 9727-N34.016 9727-N34.032 9727-N34.128 9727-N34.5 9727-N35.016 9727-N35.032 9727-N35.128 9727-N35.5 9727-N36.016 9727-N36.032 9727-N36.128 9727-N36.5 9727-N37.016 9727-N37.032 9727-N37.128 9727-N37.5 9727-N16.016 9727-N33.032 9727-N33.128 9727-N33.5 9727-N09.016 9727-N09.032 9727-N30.032 9727-N30.128 9727-N30.5 9727-N14.016 9727-N14.032 9727-N14.128 9727-N14.5
10021-472EA 76.31 USD
10021-472 10021-474 10021-476 10021-478 10021-480 10021-482 10021-484 10021-486 10021-488 10021-490 10021-492 10021-494 10021-496 10021-498 10021-500 10021-502 10021-504 10021-506 10021-508 10021-510 10021-512 10021-514 10021-516 10021-518 10021-520 10021-522 10021-524 10021-526 10021-528 10021-530 10021-532 10021-534 10021-536 10021-538 10021-540 10021-542 10021-544 10021-546 10021-548 10021-550 10021-552 10021-554 10021-556 10021-558 10021-560 10021-562 10021-564 10021-566 10021-568 10021-570 10021-572 10021-574 10021-576 10021-578 10021-580 10021-582 10021-584 10021-586 10021-588 10021-590 10021-592 10021-594 10021-596 10021-598 10021-600 10021-602 10021-604 10021-606 10021-608 10021-610 10021-612 10021-614 10021-616 10021-618 10021-620 10021-622 10021-624 10021-626 10021-628 10021-630 10021-632 10021-634 10021-636 10021-638 10021-640 10021-642 10021-644 10021-646 10021-648 10021-650 10021-652 10021-654 10021-656 10021-658 10021-660 10021-662 10021-664 10021-666 10021-668 10021-670 10021-672 10021-674 10021-676 10021-678 10021-680 10021-682 10021-684 10021-994 10021-996 10021-998 10022-000 10022-002 10022-056 10022-058 10022-060 10022-062 10022-064 10022-066 10022-114 10022-116 10022-118 10022-120
CANNON® CL Low Temperature Viscosity Standards, Cannon Instruments
Standards Viscometry Standards
CANNON® CL standards provide certified dynamic viscosity data at temperatures from –5 °C to –35 °C. These standards are used to calibrate the cold-cranking simulator for oil testing.
Certifications: Certified to ASTM D 5293 and SAE Specification J300.