General Purpose Viscosity Standards, Cannon Instruments
Supplier: CANNON INSTRUMENT CO
9727-C45.016 9727-C30.016 9727-C32.016 9727-C10.016 9727-C15.016 9727-C20.016 9727-C22.016 9727-C25.016 9727-C25.032 9727-C25.128 9727-C25.5 9727-C27.016 9727-C27.032 9727-C27.128 9727-C27.5 9727-C30.016 9727-C30.032 9727-C30.128 9727-C30.5 9727-C31.016 9727-C31.032 9727-C31.128 9727-C34.5 9727-C35.016 9727-C35.032 9727-C35.128 9727-C35.5 9727-C36.016 9727-C36.032 9727-C36.128 9727-C36.5 9727-C37.016 9727-C37.032 9727-C37.128 9727-C37.5 9727-C38.016 9727-C38.032 9727-C38.128 9727-C38.5 9727-C40.016 9727-C40.032 9727-C41.016 9727-C42.5 9727-C43.016 9727-C43.032 9727-C43.128 9727-C43.5 9727-C45.016 9727-C45.032 9727-C45.128 9727-C45.5 9727-C46.016 9727-C46.032 9727-C46.128 9727-C46.5 9727-C47.016 9727-C47.032 9727-C47.128 9727-C47.5 9727-C48.016 9727-C48.032 9727-C48.128 9727-C48.5 9727-C50.016 9727-C50.032 9727-C50.128 9727-C50.5 9727-C51.016 9727-C51.032 9727-C51.128 9727-C51.5 9727-C52.016 9727-C52.032 9727-C52.128 9727-C42.016 9727-C42.032 9727-C42.128
47735-114EA 198 USD
47735-114 47735-102 47735-104 47735-118 10021-118 10021-120 10021-122 10021-124 10021-126 10021-134 10021-136 10021-138 10021-140 10021-142 10021-144 10021-146 10021-148 10021-150 10021-152 10021-154 10021-156 10021-158 10021-160 10021-162 10021-164 10021-166 10021-168 10021-170 10021-172 10021-174 10021-176 10021-178 10021-180 10021-182 10021-184 10021-186 10021-188 10021-190 10021-192 10021-194 10021-196 10021-198 10021-200 10021-202 10021-204 10021-206 10021-208 10021-210 10021-212 10021-214 10021-216 10021-218 10021-220 10021-222 10021-224 10021-226 10021-228 10021-230 10021-232 10021-234 10021-236 10021-238 10021-240 10021-242 10021-244 10021-246 10021-248 10021-250 10021-252 10021-254 10021-256 10021-258 10021-260 10021-262 10021-264 10021-266 10021-268 10021-270 10021-272 10021-274 10021-276 10021-278 10021-280 10021-282 10022-026 10022-028 10022-030
General Purpose Viscosity Standards, Cannon Instruments
Standards Viscometry Standards
These certified standards are useful for calibration and verification procedures.
  • It is suitable for all types of viscometers
  • It also helps ensures the reliability of measurements
Certifications: Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.