Potassium nitrate ≥99.999% (K basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
19-5400-25G
19-5400-100G
100207-998EA
176.92
USD
100207-998
100209-762
Potassium nitrate ≥99.999% (K basis), PURATREM
Potassium nitrate
Formula:
KNO₃ Boiling Pt: 400 °C (dec.) Melting Pt: 334 °C Density: 2.109 Storage Temperature: Ambient |
MDL Number:
MFCD00011409 CAS Number: 7757-79-1 EINECS: 231-818-8 UN: 1486 ADR: 5.1,III REACH: 01-2119488224-35 Merck Index: 13,07733 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...