(S)-(+)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-di-t-butyl-1,1'-bi-2-naphthol dipotassium salt

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Potassium (S)-3,3'-di-tert-butyl-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-bis(olate)

19-1601-500MG 19-1601-100MG
100209-756EA 227.88 USD
100209-756 100209-754
(S)-(+)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-di-t-butyl-1,1'-bi-2-naphthol dipotassium salt
(S)-(+)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-di-t-butyl-1,1'-bi-2-naphthol dipotassium salt
Formula: C₂₈H₃₆O₂.2K
MDL Number: MFCD03788939
CAS Number: 821793-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR