(R)-(-)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-di-t-butyl-1,1'-bi-2-naphthol, dipotassium salt

Supplier: STREM CHEMICALS INC MS
Warning

19-1600-500MG 19-1600-100MG
100209-752EA 227.88 USD
100209-752 100209-750
(R)-(-)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-di-t-butyl-1,1'-bi-2-naphthol, dipotassium salt
(R)-(-)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-di-t-butyl-1,1'-bi-2-naphthol, dipotassium salt
Formula: C₂₈H₃₆O₂.2K
MDL Number: MFCD03788939
CAS Number: 350683-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR