1H,1H,2H,2H-Tridecafluoro-1-octanol ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-ol, 1H,1H,2H,2H-Perfluoro-1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol, 1H,1H,2H,2H-Perfluorooctan-1-ol, 1H,1H,2H,2H-Perfluorooctanol
09-6147-5G
09-6147-25G
100197-354EA
70.77
USD
100197-354
100209-624
1H,1H,2H,2H-Tridecafluoro-1-octanol ≥97%
1H,1H,2H,2H-Tridecafluoro-1-octanol
Organo Fluorine
Formula:
C₈H₅F₁₃O MW: 364.11 g/mol Boiling Pt: 91…93 °C (36 mmHg) Density: 1.651 |
MDL Number:
MFCD00042143 CAS Number: 647-42-7 EINECS: 211-477-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...