Pentafluoropropionic anhydride ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

09-5750-25G 09-5750-5G
100209-614EA 352.43 USD
100209-614 100196-010
Pentafluoropropionic anhydride ≥97%
Pentafluoropropionic anhydride

Organo Fluorine

Formula: C₆F₁₀O₃
Boiling Pt: 73…74 °C
Density: 1.571
Storage Temperature: Ambient
MDL Number: MFCD00000429
CAS Number: 356-42-3
EINECS: 206-604-2
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR