Bathophenanthroline ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

07-0472-1G 07-0472-250MG
100198-500EA 118.89 USD
100198-500 100209-532
Bathophenanthroline ≥99%
Bathophenanthroline
Formula: C₂₄H₁₆N₂
Boiling Pt: 508 °C (1013 hPa)
Melting Pt: 218 °C
Storage Temperature: Ambient
MDL Number: MFCD00004976
CAS Number: 1662-01-7
EINECS: 216-767-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR