Palladium Kit
Supplier: STREM CHEMICALS INC MS
96-4650-1KIT
100209-468EA 1191.75 USD
100209-468
Palladium Kit
Palladium Kit
Catalyst kit for palladium chemistry in organic synthesis
This Kit Contains the following Strem Product #s:
46-0100 Allylpalladium chloride dimer, min. 98% (500mg)
46-0400 Dichlorobis(benzonitrile)palladium(II), 99% (1g)
46-1780 Palladium(II) acetate, min. 98% (99.9+%-Pd) (1g)
46-1850 Palladium(II) chloride (99.9%-Pd) (1g)
46-2150 Tetrakis(triphenylphosphine)palladium(0), 99% (99.9+%-Pd) (5g)
46-3000 Tris(dibenzylideneacetone)dipalladium(0) (5g)
46-3010 Tris(dibenzylideneacetone)dipalladium(0) chloroform adduct (500mg)