Phosphine Ligand Kit for Palladium-Catalyzed Carbon-Carbon and Carbon-Heteroatom Bond Formation
Supplier: STREM CHEMICALS INC MS
96-1650-1KIT
100209-466EA 679.38 USD
100209-466
Phosphine Ligand Kit for Palladium-Catalyzed Carbon-Carbon and Carbon-Heteroatom Bond Formation
Phosphine Ligand Kit for Palladium-Catalyzed Carbon-Carbon and Carbon-Heteroatom Bond Formation
Phosphine; This kit is for Palladium-catalyzed carbon-carbon and carbon-heteroatom bond formation.
This Kit Contains the following Strem Product #s: 26-0270 (1g), 15-0433 (1g), 15-0380 (5g), 15-1023 (1g), 15-1045 (500mg), 15-6000 (1g), 15-6372 (500mg), 15-7720 (10g), 15-8050 (5g).
Please check strem.com for any updates and changes.