Bismuth(III) iodide ≥99.999% (Bi basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Bismuth triiodide
93-8317-5G
93-8317-25G
100197-974EA
133.05
USD
100197-974
100209-464
Bismuth(III) iodide ≥99.999% (Bi basis), PURATREM
Bismuth (III) iodide
Formula:
BiI₃ Boiling Pt: ~500 °C Melting Pt: 408 °C Density: 5.778 |
MDL Number:
MFCD00010894 CAS Number: 7787-64-6 EINECS: 232-127-4 UN: 1759 ADR: 8,III Merck Index: 12,01308 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...