Protoporphyrin IX

Supplier: STREM CHEMICALS INC MS

Synonyms: Ooporphyrin, Kammerer’s porphyrin, 3,7,12,17-Tetramethyl-8,13-divinyl-2,18-porphinedipropionic acid

07-1820-1G
100209-296EA 384.98 USD
100209-296
Protoporphyrin IX
Protoporphyrin IX
Formula: C₃₄H₃₄N₄O₄
MDL Number: MFCD00151109
CAS Number: 553-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR