Lithium-tert-butanolate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

03-0780-100G 03-0780-25G
100209-286EA 396.31 USD
100209-286 100199-686
Lithium-tert-butanolate ≥98%
Lithium tert-butanolate

Alkoxide, Volatile Precursors for CVD

Formula: C₄H₉LiO
Density: 0.89
MDL Number: MFCD00050479
CAS Number: 1907-33-1
UN: 3206
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR