Ruthenium(III) chloride hydrate ≥99.9% (Ru basis) (40-43% Ru)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate

44-5880-5G 44-5880-25G 44-5880-1G
100201-660EA 263.26 USD
100201-660 100209-262 100206-664
Ruthenium(III) chloride hydrate ≥99.9% (Ru basis) (40-43% Ru)
Ruthenium (III) chloride hydrate
Formula: Cl₃Ru·xH₂O
Melting Pt: 100 °C (dec.)
Density: 3.11 g/cm³ (20 °C)
MDL Number: MFCD00149844
CAS Number: 14898-67-0
EINECS: 233-167-5
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR